COMH93D61A8E Combi-Blocks, Inc.

1,1,3,3,6-Pentamethyl-7-nitro-5-indanamine

  • MDL number MFCD03783538
  • Cas Number : 255393-53-4
  • Molecular Weight : 248.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-3232

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-3232
Availability details for
Quantity
  • Availability
  • Add to Cart