COMH93D61AC8 Combi-Blocks, Inc.

Ethyl 4-methyl-2-(4-methyl-1,4-diazepan-1-yl)-1,3-thiazole-5-carboxylate

  • MDL number MFCD31693753
  • Cas Number : 2107633-95-2
  • Molecular Weight : 283.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-3296

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-3296
Availability details for
Quantity
  • Availability
  • Add to Cart