COMH93D61B0D Combi-Blocks, Inc.

3-Fluoro-5-phenylbenzaldehyde

  • MDL number MFCD31716312
  • Cas Number : 1645949-36-5
  • Molecular Weight : 200.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-3374

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-3374
Availability details for
Quantity
  • Availability
  • Add to Cart