COMH93D61B10 Combi-Blocks, Inc.

Narcissoside

  • MDL number MFCD00017734
  • Cas Number : 604-80-8
  • Molecular Weight : 624.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-3379

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-3379
Availability details for
Quantity
  • Availability
  • Add to Cart