COMH93D61B2B Combi-Blocks, Inc.

4-(3-Cyclopropylmethoxy-phenoxy)-piperidine

  • MDL number MFCD31716350
  • Cas Number : 1936450-06-4
  • Molecular Weight : 247.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-3410

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-3410
Availability details for
Quantity
  • Availability
  • Add to Cart