COMH93D61B71 Combi-Blocks, Inc.

3-[(3-Chloro-4-fluorophenoxy)methyl]azetidine

  • MDL number MFCD31700266
  • Cas Number : 1864587-95-0
  • Molecular Weight : 215.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-3499

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-3499
Availability details for
Quantity
  • Availability
  • Add to Cart