COMH93D61B7D Combi-Blocks, Inc.

1-(5-Bromoindolin-1-yl)-2-(dimethylamino)ethanone

  • MDL number MFCD31725235
  • Cas Number : 1350377-17-1
  • Molecular Weight : 283.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-3514

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-3514
Availability details for
Quantity
  • Availability
  • Add to Cart