COMH93D61BA9 Combi-Blocks, Inc.

N-Cyclopropyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzenesulfonamide

  • MDL number MFCD32198945
  • Molecular Weight : 323.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-3566

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-3566
Availability details for
Quantity
  • Availability
  • Add to Cart