COMH93D61D5E Combi-Blocks, Inc.

2-[4-Chloro-5-cyclopropyl-3-(difluoromethyl)-1h-pyrazol-1-yl]acetohydrazide

  • MDL number MFCD04969906
  • Cas Number : 1004643-39-3
  • Molecular Weight : 264.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-4089

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-4089
Availability details for
Quantity
  • Availability
  • Add to Cart