COMH93D61DBA Combi-Blocks, Inc.

2-[3,5-Bis(difluoromethyl)-1h-pyrazol-1-yl]acetohydrazide

  • MDL number MFCD03419713
  • Cas Number : 1001518-87-1
  • Molecular Weight : 240.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-4204

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-4204
Availability details for
Quantity
  • Availability
  • Add to Cart