COMH93D61DCA Combi-Blocks, Inc.

N,N'-Bis(2-chloroethyl)-1h-imidazole-4,5-dicarboxamide

  • MDL number MFCD31693804
  • Cas Number : 4787-07-9
  • Molecular Weight : 279.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-4222

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-4222
Availability details for
Quantity
  • Availability
  • Add to Cart