COMH93D61DE7 Combi-Blocks, Inc.

N-(Morpholin-2-ylmethyl)-2,4-dioxo-1h-pyrimidine-6-carboxamide hydrochloride

  • MDL number MFCD31926084
  • Molecular Weight : 290.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-4256

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-4256
Availability details for
Quantity
  • Availability
  • Add to Cart