COMH93D61EB9 Combi-Blocks, Inc.

N-Cyclopropyl-3-hydroxy-4-iodobenzamide

  • MDL number MFCD31735215
  • Cas Number : 1691826-00-2
  • Molecular Weight : 303.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-4511

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-4511
Availability details for
Quantity
  • Availability
  • Add to Cart