COMH93D61EC6 Combi-Blocks, Inc.

1-(2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)urea

  • MDL number MFCD31746796
  • Cas Number : 2246863-66-9
  • Molecular Weight : 276.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-4530

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-4530
Availability details for
Quantity
  • Availability
  • Add to Cart