COMH93D61F26 Combi-Blocks, Inc.

N-(2-Methoxy-6-nitrophenyl)prop-2-enamide

  • MDL number MFCD31924894
  • Cas Number : 2095669-62-6
  • Molecular Weight : 222.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-4647

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-4647
Availability details for
Quantity
  • Availability
  • Add to Cart