COMH93D61F45 Combi-Blocks, Inc.

4-Bromo-2-chloro-n-(3-sulfamoylphenyl)benzenesulfonamide

  • MDL number MFCD31716583
  • Molecular Weight : 425.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-4684

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-4684
Availability details for
Quantity
  • Availability
  • Add to Cart