COMH93D61F80 Combi-Blocks, Inc.

3-Chloro-4'-ethynyl-biphenyl

  • MDL number MFCD31716492
  • Cas Number : 2279122-69-7
  • Molecular Weight : 212.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-4757

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-4757
Availability details for
Quantity
  • Availability
  • Add to Cart