COMH93D61F96 Combi-Blocks, Inc.

4-((3-Bromo-4-methylphenoxy)methyl)-2,2-dimethyl-1,3-dioxolane

  • MDL number MFCD32198840
  • Molecular Weight : 301.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-4781

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-4781
Availability details for
Quantity
  • Availability
  • Add to Cart