COMH93D61FC3 Combi-Blocks, Inc.

3-(3-Fluorophenyl)pyridine-2,6-diamine

  • MDL number MFCD32067034
  • Molecular Weight : 203.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-4830

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-4830
Availability details for
Quantity
  • Availability
  • Add to Cart