COMH93D62021 Combi-Blocks, Inc.

Dihydroxybis(hydrogen lactato)titanium(iv)

  • MDL number MFCD32062935
  • Cas Number : 79110-90-0
  • Molecular Weight : 260.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-4938

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-4938
Availability details for
Quantity
  • Availability
  • Add to Cart