COMH93D6218C Combi-Blocks, Inc.

4-Hydrazino-5,6,7,8,9,10-hexahydrocycloocta[4,5]thieno[2,3-d]pyrimidine

  • MDL number MFCD31693751
  • Cas Number : 40106-64-7
  • Molecular Weight : 248.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-5376

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-5376
Availability details for
Quantity
  • Availability
  • Add to Cart