COMH93D62194 Combi-Blocks, Inc.

N-Isopropyl-4-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-2-trifluoromethoxy-benzamide

  • MDL number MFCD31760062
  • Molecular Weight : 373.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-5385

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-5385
Availability details for
Quantity
  • Availability
  • Add to Cart