COMH93D621D3 Combi-Blocks, Inc.

6-[(tert-Butoxy)carbonyl]-2-oxa-6-azaspiro[3.3]heptane-5-carboxylic acid

  • MDL number MFCD31728994
  • Cas Number : 2168618-83-3
  • Molecular Weight : 243.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-5467

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-5467
Availability details for
Quantity
  • Availability
  • Add to Cart