COMH93D622FA Combi-Blocks, Inc.

1-Benzyl-3-(((tert-butyldimethylsilyl)oxy)methyl)tetrahydrofuro[3,4-c]isoxazol-6(3h)-one

  • MDL number MFCD31803601
  • Cas Number : 2205333-31-7
  • Molecular Weight : 363.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-5827

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-5827
Availability details for
Quantity
  • Availability
  • Add to Cart