COMH93D62334 Combi-Blocks, Inc.

4-((3-((tert-Butyldimethylsilyl)oxy)phenyl)(hydroxy)methyl)-n,n-diethylbenzamide

  • MDL number MFCD32196767
  • Cas Number : 155765-75-6
  • Molecular Weight : 413.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-5899

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-5899
Availability details for
Quantity
  • Availability
  • Add to Cart