COMH93D62340 Combi-Blocks, Inc.

3-Amino-n-methyl-4-trifluoromethoxy-benzamide

  • MDL number MFCD32198778
  • Molecular Weight : 234.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-5913

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-5913
Availability details for
Quantity
  • Availability
  • Add to Cart