COMH93D62359 Combi-Blocks, Inc.

1-(3-Bromo-5-trifluoromethyl-phenoxymethyl)-cyclopropylamine hydrochloride

  • MDL number MFCD32198747
  • Molecular Weight : 346.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-5942

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-5942
Availability details for
Quantity
  • Availability
  • Add to Cart