COMH93D6239F Combi-Blocks, Inc.

1-Bromo-2-ethynyl-4-isopropoxy-benzene

  • MDL number MFCD31716662
  • Cas Number : 2279124-00-2
  • Molecular Weight : 239.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-6023

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-6023
Availability details for
Quantity
  • Availability
  • Add to Cart