COMH93D623A0 Combi-Blocks, Inc.

Fcla

  • MDL number MFCD04037231
  • Cas Number : 133118-06-6
  • Molecular Weight : 675.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-6024

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-6024
Availability details for
Quantity
  • Availability
  • Add to Cart