COMH93D623C8 Combi-Blocks, Inc.

3,3-Difluoro-1-((3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)sulfonyl)pyrrolidine

  • MDL number MFCD32198948
  • Molecular Weight : 373.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-6073

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-6073
Availability details for
Quantity
  • Availability
  • Add to Cart