COMH93D623CA Combi-Blocks, Inc.

tert-Butyl 3-((2-fluoro-4-formylphenoxy)methyl)azetidine-1-carboxylate

  • MDL number MFCD31916061
  • Cas Number : 2340293-90-3
  • Molecular Weight : 309.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-6075

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-6075
Availability details for
Quantity
  • Availability
  • Add to Cart