COMH93D6252F Combi-Blocks, Inc.

tert-Butyl n-[(1s,2s,4s)-rel-4-amino-2-fluoro-cyclohexyl]carbamate hydrochloride

  • MDL number MFCD31942843
  • Molecular Weight : 268.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-6506

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-6506
Availability details for
Quantity
  • Availability
  • Add to Cart