COMH93D625D3 Combi-Blocks, Inc.

1-(Benzyloxy)-4-ethynyl-2-fluorobenzene

  • MDL number MFCD31916064
  • Cas Number : 722537-46-4
  • Molecular Weight : 226.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-6698

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-6698
Availability details for
Quantity
  • Availability
  • Add to Cart