COMH93D625F8 Combi-Blocks, Inc.

4-Bromo-6,7-dimethoxy-indan-1-one

  • MDL number MFCD00088977
  • Cas Number : 18028-29-0
  • Molecular Weight : 271.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-6745

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-6745
Availability details for
Quantity
  • Availability
  • Add to Cart