COMH93D62645 Combi-Blocks, Inc.

4'-Ethynyl-3,5-difluoro-biphenyl

  • MDL number MFCD31716494
  • Cas Number : 2279123-83-8
  • Molecular Weight : 214.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-6844

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-6844
Availability details for
Quantity
  • Availability
  • Add to Cart