COMH93D62660 Combi-Blocks, Inc.

3-((Hexyloxy)methyl)heptane

  • MDL number MFCD31746799
  • Molecular Weight : 214.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-6875

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-6875
Availability details for
Quantity
  • Availability
  • Add to Cart