COMH93D62666 Combi-Blocks, Inc.

[1-(4-Bromo-2-chloro-phenoxymethyl)-cyclopropyl]-carbamic acid tert-butyl ester

  • MDL number MFCD32198734
  • Molecular Weight : 376.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-6882

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-6882
Availability details for
Quantity
  • Availability
  • Add to Cart