COMH93D62766 Combi-Blocks, Inc.

(2S,5R)-5-Methyl-2-(trifluoromethyl)morpholine4-methylbenzenesulfonate

  • MDL number MFCD31803324
  • Molecular Weight : 341.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-7187

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-7187
Availability details for
Quantity
  • Availability
  • Add to Cart