COMH93D62780 Combi-Blocks, Inc.

N-2-Adamantyl-3-oxobutanamide

  • MDL number MFCD02215652
  • Cas Number : 63664-40-4
  • Molecular Weight : 235.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-7222

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-7222
Availability details for
Quantity
  • Availability
  • Add to Cart