COMH93D6278A Combi-Blocks, Inc.

Sertindole

  • MDL number MFCD00867749
  • Cas Number : 106516-24-9
  • Molecular Weight : 440.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-7235

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-7235
Availability details for
Quantity
  • Availability
  • Add to Cart