COMH93D62790 Combi-Blocks, Inc.

Isopropyltrichlorosilane

  • MDL number MFCD01861719
  • Cas Number : 4170-46-1
  • Molecular Weight : 177.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-7245

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-7245
Availability details for
Quantity
  • Availability
  • Add to Cart