COMH93D627B4 Combi-Blocks, Inc.

7H-Pyrrolo[2,3-d]pyrimidin-4-ylmethanamine dihydrochloride

  • MDL number MFCD31689238
  • Molecular Weight : 221.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-7291

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-7291
Availability details for
Quantity
  • Availability
  • Add to Cart