COMH93D627CE Combi-Blocks, Inc.

([3-Chloro-4-(trifluoromethoxy)phenyl]methyl)(methyl)amine

  • MDL number MFCD31760083
  • Cas Number : 2229280-88-8
  • Molecular Weight : 239.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-7325

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-7325
Availability details for
Quantity
  • Availability
  • Add to Cart