COMH93D627DC Combi-Blocks, Inc.

2,3-Dicyanohydroquinone hydrate

  • MDL number MFCD31811642
  • Molecular Weight : 178.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-7347

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-7347
Availability details for
Quantity
  • Availability
  • Add to Cart