COMH93D62849 Combi-Blocks, Inc.

Methyl 4-[(chloroacetyl)(methyl)amino]benzoate

  • MDL number MFCD31806068
  • Cas Number : 99105-47-2
  • Molecular Weight : 241.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-7483

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-7483
Availability details for
Quantity
  • Availability
  • Add to Cart