COMH93D62A0A Combi-Blocks, Inc.

Methyl-[2-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-5-trifluoromethyl-benzyl]-carbamic acid benzyl ester

  • MDL number MFCD31760268
  • Molecular Weight : 449.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-8036

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-8036
Availability details for
Quantity
  • Availability
  • Add to Cart