COMH93D62A80 Combi-Blocks, Inc.

1-(4-Chlorophenyl)-2-(1h-imidazol-1-yl)-1-ethanone

  • MDL number MFCD00215352
  • Cas Number : 24155-32-6
  • Molecular Weight : 220.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-8174

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-8174
Availability details for
Quantity
  • Availability
  • Add to Cart