COMH93D62A82 Combi-Blocks, Inc.

N-(2-Bromo-4-fluorobenzyl)-1-cyclopropyl-n-methylmethanamine hydrochloride

  • MDL number MFCD31924906
  • Cas Number : 2358751-60-5
  • Molecular Weight : 308.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-8177

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-8177
Availability details for
Quantity
  • Availability
  • Add to Cart