COMH93D62AA6 Combi-Blocks, Inc.

tert-Butyl (7-bromoisoquinolin-3-yl)carbamate

  • MDL number MFCD30188704
  • Cas Number : 2089301-09-5
  • Molecular Weight : 323.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-8225

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-8225
Availability details for
Quantity
  • Availability
  • Add to Cart