COMH93D62AC0 Combi-Blocks, Inc.

6-Chloro-4-thiophen-2-yl-pyridazin-3-ylamine

  • MDL number MFCD31700281
  • Cas Number : 1903632-40-5
  • Molecular Weight : 211.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-8252

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-8252
Availability details for
Quantity
  • Availability
  • Add to Cart